ENA358812943 Enamine

3-amino-3-(2-methylphenyl)propanoic acid

  • MDL number MFCD01871311
  • Cas Number : 68208-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68315

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68315
Availability details for
Quantity
  • Availability
  • Add to Cart