ENA358812949 Enamine

4-[(1-methyl-1H-imidazol-2-yl)methyl]piperidine

  • MDL number MFCD18381588
  • Cas Number : 1316220-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71120
Availability details for
Quantity
  • Availability
  • Add to Cart