ENA358812950 Enamine

3-(dimethyl-1H-1,2,4-triazol-1-yl)piperidine

  • MDL number MFCD14632028
  • Cas Number : 1249679-72-8
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3,5-dimethyl-1H-1,2,4-triazol-1-yl)piperidine
Inchi Code 1S/C9H16N4/c1-7-11-8(2)13(12-7)9-4-3-5-10-6-9/h9-10H,3-6H2,1-2H3
Inchi Key BAOHPMOOLLSPSO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71195
Availability details for
Quantity
  • Availability
  • Add to Cart