ENA358812966 Enamine

thiolane-2-carboxylic acid

  • MDL number MFCD11520782
  • Cas Number : 19418-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48...50
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77924

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77924
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart