ENA358812977 Enamine

1-(bromomethyl)-2-fluoro-4-nitrobenzene

  • MDL number MFCD10000853
  • Cas Number : 127349-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84033
Availability details for
Quantity
  • Availability
  • Add to Cart