ENA358812981 Enamine

2-(2,5-dimethoxyphenyl)-2,2-difluoroacetic acid

  • MDL number MFCD17223386
  • Cas Number : 1342352-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84979
Availability details for
Quantity
  • Availability
  • Add to Cart