ENA358812983 Enamine

5-(chloromethyl)-3-(2-phenylethyl)-1,2,4-oxadiazole

  • MDL number MFCD14654387
  • Cas Number : 1247725-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85104

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85104
Availability details for
Quantity
  • Availability
  • Add to Cart