ENA358813003 Enamine

1,1,1-trifluoro-2-methylbutan-2-ol

  • MDL number MFCD16067935
  • Cas Number : 507-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90705
Availability details for
Quantity
  • Availability
  • Add to Cart