ENA358813005 Enamine

2-methyl-6-(trifluoromethyl)morpholine

  • MDL number MFCD19603109
  • Cas Number : 1342385-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90726
Availability details for
Quantity
  • Availability
  • Add to Cart