ENA358813010 Enamine

5-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11226745
  • Cas Number : 1402447-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90915
Availability details for
Quantity
  • Availability
  • Add to Cart