ENA358813036 Enamine

4-(quinoxalin-2-yl)phenol

  • MDL number MFCD00117343
  • Cas Number : 33707-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-94827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94827
Availability details for
Quantity
  • Availability
  • Add to Cart