ENA358813038 Enamine

ethyl 3-hydroxy-2-methylpropanoate

  • MDL number MFCD16087525
  • Cas Number : 89534-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95131
Availability details for
Quantity
  • Availability
  • Add to Cart