ENA358813055 Enamine

[2-(4-methoxyphenyl)-1,3-thiazol-4-yl]methanamine

  • MDL number MFCD06738418
  • Cas Number : 857997-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95693
Availability details for
Quantity
  • Availability
  • Add to Cart