ENA358813057 Enamine

4,6-dichloro-2,3-dihydro-1-benzofuran-3-one

  • MDL number MFCD12093558
  • Cas Number : 857062-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95781
Availability details for
Quantity
  • Availability
  • Add to Cart