ENA358813065 Enamine

3-(thiophen-2-yl)thiomorpholine

  • MDL number MFCD16302499
  • Cas Number : 933725-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95871
Availability details for
Quantity
  • Availability
  • Add to Cart