ENA358813084 Enamine

7-bromo-6-chloro-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD11104470
  • Cas Number : 5791-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 281...283
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96049
Availability details for
Quantity
  • Availability
  • Add to Cart