ENA358813085 Enamine

3-amino-4,5-dimethoxybenzoic acid

  • MDL number MFCD17276595
  • Cas Number : 861556-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96050

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96050
Availability details for
Quantity
  • Availability
  • Add to Cart