ENA358813092 Enamine

5-chloro-2,3-dihydro-1H-inden-2-one

  • MDL number MFCD09744099
  • Cas Number : 74444-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-75
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96075
Availability details for
Quantity
  • Availability
  • Add to Cart