ENA358813099 Enamine

4-methyl-1,4-diazaspiro[5.5]undecane

  • MDL number MFCD20427314
  • Cas Number : 933689-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96090
Availability details for
Quantity
  • Availability
  • Add to Cart