ENA358813106 Enamine

2-(5-phenyl-1,3,4-oxadiazol-2-yl)ethan-1-amine

  • MDL number MFCD09883767
  • Cas Number : 933734-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96407

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96407
Availability details for
Quantity
  • Availability
  • Add to Cart