ENA358813110 Enamine

3-methyl-4-(trifluoromethanesulfonamido)benzoic acid

  • MDL number MFCD11550613
  • Cas Number : 1094630-91-7
  • Molecular Weight : 283.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96513
Availability details for
Quantity
  • Availability
  • Add to Cart