ENA358813111 Enamine

2-(piperidin-3-yl)acetamide

  • MDL number MFCD16839212
  • Cas Number : 196613-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96530
Availability details for
Quantity
  • Availability
  • Add to Cart