ENA358813123 Enamine

3-phenylbutan-2-one

  • MDL number MFCD00156648
  • Cas Number : 769-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96836

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96836
Availability details for
Quantity
  • Availability
  • Add to Cart