ENA358813125 Enamine

methyl 3-(4-bromo-1H-pyrazol-1-yl)-2-methylpropanoate

  • MDL number MFCD04967354
  • Cas Number : 1005615-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96855
Availability details for
Quantity
  • Availability
  • Add to Cart