ENA358813127 Enamine

2-(hydroxymethyl)-5-phenyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD07759276
  • Cas Number : 900449-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96857
Availability details for
Quantity
  • Availability
  • Add to Cart