ENA358813128 Enamine

methyl 3-amino-5-chloro-2-fluorobenzoate

  • MDL number MFCD17276537
  • Cas Number : 1269421-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96858
Availability details for
Quantity
  • Availability
  • Add to Cart