ENA358813145 Enamine

5-ethylimidazolidine-2,4-dione

  • MDL number MFCD00059161
  • Cas Number : 15414-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96932

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96932
Availability details for
Quantity
  • Availability
  • Add to Cart