ENA358813153 Enamine

4-(4-aminophenyl)-4-oxobutanoic acid

  • MDL number MFCD12196376
  • Cas Number : 6945-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97000
Availability details for
Quantity
  • Availability
  • Add to Cart