ENA358813154 Enamine

4-methoxyquinoline-3-carboxylic acid

  • MDL number MFCD14632644
  • Cas Number : 74557-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97002
Availability details for
Quantity
  • Availability
  • Add to Cart