ENA358813156 Enamine

2-{4-[(benzyloxy)carbonyl]piperazin-1-yl}acetic acid

  • MDL number MFCD13720327
  • Cas Number : 1153907-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97006
Availability details for
Quantity
  • Availability
  • Add to Cart