ENA358813168 Enamine

1-[(propan-2-yloxy)carbonyl]-3-(trifluoromethyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD19000634
  • Cas Number : 1365836-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97065
Availability details for
Quantity
  • Availability
  • Add to Cart