ENA358813171 Enamine

1-[4-(propan-2-yl)phenyl]propan-1-amine

  • MDL number MFCD03419830
  • Cas Number : 832741-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97094
Availability details for
Quantity
  • Availability
  • Add to Cart