ENA358813173 Enamine

1-[4-(3,4-difluorophenoxy)pyridin-2-yl]methanamine

  • MDL number MFCD17136249
  • Cas Number : 1365836-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97167
Availability details for
Quantity
  • Availability
  • Add to Cart