ENA358813178 Enamine

pyrrolidine-2-thione

  • MDL number MFCD00049351
  • Cas Number : 2295-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97179

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-97179
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart