ENA358813180 Enamine

1-(2-methoxyphenyl)-3-methylbutan-1-amine

  • MDL number MFCD11935358
  • Cas Number : 1181632-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97182
Availability details for
Quantity
  • Availability
  • Add to Cart