ENA358813182 Enamine

3-ethynyl-5-fluoropyridine

  • MDL number MFCD13175288
  • Cas Number : 872122-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97184
Availability details for
Quantity
  • Availability
  • Add to Cart