ENA358813184 Enamine

chloromethyl benzoate

  • MDL number MFCD00040116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-97187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97187
Availability details for
Quantity
  • Availability
  • Add to Cart