ENA358813192 Enamine

5-bromo-N-(1,2,3,4-tetrahydroquinolin-6-yl)thiophene-2-carboxamide

  • MDL number MFCD18557427
  • Cas Number : 1375473-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97240
Availability details for
Quantity
  • Availability
  • Add to Cart