ENA358813196 Enamine

N-cyclopropyl-7,8-dimethyl-2,3,4,5-tetrahydro-1-benzoxepin-5-amine

  • MDL number MFCD12423570
  • Cas Number : 1153408-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97246
Availability details for
Quantity
  • Availability
  • Add to Cart