ENA358813197 Enamine

1-(4-methylbenzenesulfonyl)piperidin-4-one

  • MDL number MFCD00218704
  • Cas Number : 33439-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97258
Availability details for
Quantity
  • Availability
  • Add to Cart