ENA358813214 Enamine

N-(2-chloroethyl)-N-methylaniline

  • MDL number MFCD01670150
  • Cas Number : 1669-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97358

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-97358
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart