ENA358813217 Enamine

1-(5-tert-butyl-1,3-oxazol-2-yl)methanamine

  • MDL number MFCD09880833
  • Cas Number : 1023814-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97364
Availability details for
Quantity
  • Availability
  • Add to Cart