ENA358813224 Enamine

2-[4-(trifluoromethyl)phenyl]cyclohexan-1-one

  • MDL number MFCD16659587
  • Cas Number : 52776-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-97386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97386
Availability details for
Quantity
  • Availability
  • Add to Cart