ENA358813236 Enamine

2-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)-4-methylaniline

  • MDL number MFCD11117014
  • Cas Number : 1036600-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97446
Availability details for
Quantity
  • Availability
  • Add to Cart