ENA358813238 Enamine

2-(1H-imidazol-1-yl)-3-methylbutanoic acid

  • MDL number MFCD08668217
  • Cas Number : 745001-73-4
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97451
Availability details for
Quantity
  • Availability
  • Add to Cart