ENA358813252 Enamine

3-(quinolin-2-yl)butan-2-ol

  • MDL number MFCD18290456
  • Cas Number : 1344310-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97538
Availability details for
Quantity
  • Availability
  • Add to Cart