ENA358813256 Enamine

2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD07375329
  • Cas Number : 762287-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97550

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97550
Availability details for
Quantity
  • Availability
  • Add to Cart