ENA358813260 Enamine

5-chloro-7-methyl-1,3-benzoxazol-2-amine

  • MDL number MFCD16742778
  • Cas Number : 1250209-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97625
Availability details for
Quantity
  • Availability
  • Add to Cart