ENA358813263 Enamine

methyl 4-(difluoromethoxy)-1H-indole-2-carboxylate

  • MDL number MFCD09042155
  • Cas Number : 926219-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97631
Availability details for
Quantity
  • Availability
  • Add to Cart