ENA358813266 Enamine

diethyl[2-(pyrrolidin-3-yloxy)ethyl]amine

  • MDL number MFCD16709454
  • Cas Number : 1247410-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97636
Availability details for
Quantity
  • Availability
  • Add to Cart